Order Entry
Canada
Orders LinkContactUsLinkComponent
Netwell™ Inserts, Sterile, Corning

Netwell™ Inserts, Sterile, Corning

6, 24 mm, 74 µm, 6/Plate
Catalog # 29442-136
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  3479
Netwell™ Inserts, Sterile, Corning
Catalog # 29442-136
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Netwell™ Inserts, Sterile, Corning
Netwell™ Inserts, Sterile, Corning

6, 24 mm, 74 µm, 6/Plate

Price
651,10 $
New Customer Promotional Price
Log in for availability
Case of 48
Product Details & Documents

Polyester mesh bottoms attached to PS rings or housing. Netwell™ inserts are used as tissue carriers, supports and strainers of small organs, tissue slices or explants at the air-media interface.

  • Handy carrier for immunocytochemical staining of tissue slices
  • Provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
  • Available in two mesh sizes and diameters
  • Supplied sterile and pre-loaded in 6- or 12-well plates
  • 24 mm Netwell inserts fit in Corning® 50 ml plastic centrifuge tubes

Polystyrene reagent trays are available in black for improved visibility of tissue sections, or white for colorimetric reaction contrast. Specially designed carriers and handles permit the simultaneous processing of up to 12 samples per carrier. Each carrier kit contains eight carriers and eight handles.

Specifications
  • No. of wells:
    6
  • Packaging:
    6/Plate
  • Mesh size:
    74 µm
  • Membrane diameter:
    24 mm
  • Cat. No.:
    29442-136
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  • Catalog #
    Membrane Diameter
    Availability
    Price
  • Carrier kit, 6-well
    Carrier kit, 6-well
    29442-158
    24 mm
    Log in for availability
    Case of 8 - $356.15

Product Information

  • Catalog #
    Availability
    Price
  • Reagent Tray, Black
    Reagent Tray, Black
    29442-152
    Log in for availability
    Case of 200 - $357.00

Product Information

  • Catalog #
    Membrane Diameter
    Availability
    Price
  • Carrier kit, 12-well
    Carrier kit, 12-well
    29442-156
    15 mm
    Log in for availability
    Case of 8 - $398.05

Product Information

  • Catalog #
    Availability
    Price
  • Reagent Tray, White
    Reagent Tray, White
    29442-154
    Log in for availability
    Case of 200 - $420.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.