Transwell® Clear Inserts, Polyester (PET) Membrane
24, 0.33 cm², 6.5 mm, Polyester, 0.4 µm, 12/Plate, 4 Plates
Supplier: Corning
Supplier #: 3470
Transwell® Clear Inserts, Polyester (PET) Membrane
Catalog # 29442-082
Supplier: Corning
Jump to:
Product Details & Documents
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
- w Produce a cell culture environment that closely resembles an in vivo state
- Co-culture cells with or without cell-to-cell contact
- Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
- Treated for optimal cell attachment
- Excellent visibility under phase contrast microscopy
- Supplied in multiple well plates
- Sterile
Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.
Documents
Certificates
Search by Lot Number3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture Quick Reference Ordering Guide Corning 3D Cell Culture.pdf Corning BioCoat, Falcon and Transwell Permeable Supports Corning Permeable Supports Selection Guide
Product Literature
More
Corning Transwell Permeable Supports.pdf
Promotion Material
COVID-19 with Lung Organoids
Application Notes
Specifications
- No. of wells:24
- Pore size:0.4 µm
- Packaging:12/Plate, 4 Plates
- Culture area:0.33 cm²
- Membrane diameter:6.5 mm
- Membrane material:Polyester
- Cat. No.:29442-082
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



