Order Entry
Canada
Orders LinkContactUsLinkComponent
Transwell® Clear Inserts, Polyester (PET) Membrane

Transwell® Clear Inserts, Polyester (PET) Membrane

6, 4.67 cm², 24 mm, Polyester, 0.4 µm, 6/Plate
Catalog # 29442-074
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  3450
Transwell® Clear Inserts, Polyester (PET) Membrane
Catalog # 29442-074
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Transwell® Clear Inserts, Polyester (PET) Membrane
Transwell® Clear Inserts, Polyester (PET) Membrane

6, 4.67 cm², 24 mm, Polyester, 0.4 µm, 6/Plate

Price
291,65 $
New Customer Promotional Price
Log in for availability
Case of 24
Product Details & Documents

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

  • w Produce a cell culture environment that closely resembles an in vivo state
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
  • Treated for optimal cell attachment
  • Excellent visibility under phase contrast microscopy
  • Supplied in multiple well plates
  • Sterile

Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally.

Specifications
  • No. of wells:
    6
  • Pore size:
    0.4 µm
  • Packaging:
    6/Plate
  • Culture area:
    4.67 cm²
  • Membrane diameter:
    24 mm
  • Membrane material:
    Polyester
  • Cat. No.:
    29442-074
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.