Nalgene® Filtering Flasks, Polypropylene, Thermo Scientific
500 ml, 7
Catalog # 29417-026
Jump to:
Discontinued
Product Details & Documents
One-piece molded for strength and stability
Withstand full vacuum to 762mm Hg (30in. Hg). Tubulation at 45° angle prevents tipping. Oval marking area. Barbed adapter accepts 6.4mm (1/4") to 9.5mm (3/8") I.D. tubing. Autoclavable.
Specifications
- Capacity:500 ml
- Rubber stopper size:7