Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
12 L, 11
Catalog # 29111-230
Supplier:   Corning
 :  4060-12L
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Catalog # 29111-230
Supplier:  Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
12 L, 11
Price
$556.00
Log in for availability
Case of 1
Product Details & Documents
Manufactured from borosilicate glass.
  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability
Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).
Specifications
  • Capacity:
    12 L
  • Stopper no.:
    11