Orders LinkContactUsLinkComponent
Conductivity Solutions, YSI
1 pint, 10,000 µmhos/cm, 1.0% at 25 °C
Catalog # 23200-280
Supplier:   YSI
 :  060911
Conductivity Solutions, YSI
Catalog # 23200-280
Supplier:  YSI
CAS Number:  
Conductivity Solutions, YSI
Conductivity Solutions, YSI
1 pint, 10,000 µmhos/cm, 1.0% at 25 °C
CAS Number:  
Price
$386.53
Log in for availability
Case of 8 (1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Use YSI Conductivity Solutions to calibrate conductivity cell and meter together as a system for maximum measurement accuracy.
Solutions are standardized against NIST Standard Reference Material potassium chloride (KCl). Each bottle includes calibration instruments and temperature correction information. Packaged in glass quart and plastic pint bottles.
Specifications
Specifications
  • Size:
    1 pint
  • Accuracy:
    1.0% at 25 °C
  • Conductivity:
    10,000 µmhos/cm
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.