Orders LinkContactUsLinkComponent
Corning® CellBIND® Flasks, Sterile, Corning | Avantor
Corning® CellBIND® Flasks, Sterile, Corning
Standard with Bar Code, 250 ml, Rectangular, 175 cm2, Angled, Vented cap
Catalog # 22250-134
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Fournisseur #:  431328
Corning® CellBIND® Flasks, Sterile, Corning
Catalog # 22250-134
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Corning® CellBIND® Flasks, Sterile, Corning
Corning® CellBIND® Flasks, Sterile, Corning
Standard with Bar Code, 250 ml, Rectangular, 175 cm2, Angled, Vented cap
Price
$1,521.50
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 84
Product Details & Documents
CellBIND® surface treatment improves cell attachment by incorporating more oxygen into the cell culture surface, rendering it more hydrophilic and increasing surface stability
Manufactured from optically clear polystyrene, all flasks undergo rigorous QC testing for consistency and reproducibility. Standard flasks are available with or without bar coding for use in automated systems.
RoboFlask™ vessels are specifically formatted to a standard microplate footprint for use in automated cell maintenance and assay systems. All flasks are sterilized by gamma radiation and certified nonpyrogenic.
CellBIND® surfaces are nonbiological and require no special handling or storage.
Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Spécifications
  • Cap type:
    Vented cap
  • Capacity:
    250 ml
  • Description:
    Standard with Bar Code
  • Shape:
    Rectangular
  • Neck style:
    Angled
  • Growth area:
    175 cm2
  • Cat. No.:
    22250-134
  • Environmental certification:
    RECs, ISO 14001 - Oneonta
  • Environmentally Preferable:
    Low Manufacturing Impact