Order Entry
Canada
Orders LinkContactUsLinkComponent
2-Needle Array™ Electrode Kits, BTX®

2-Needle Array™ Electrode Kits, BTX®

2-Needle Array™, electrode gap 10 mm, 8 cm, 20 mm
Catalog # 22221-522
Supplier:   Harvard Apparatus
Supplier #:  45-0167
2-Needle Array™ Electrode Kits, BTX®
Catalog # 22221-522
Supplier:  Harvard Apparatus
2-Needle Array™ Electrode Kits, BTX®
2-Needle Array™ Electrode Kits, BTX®

2-Needle Array™, electrode gap 10 mm, 8 cm, 20 mm

Price
$1,122.24
Log in for availability
Each
Product Details & Documents

In vivo-style electrodes are designed specifically for muscle gene delivery

Kit 22221-522 is recommended for larger muscle masses, such as rat gastrocnemus. Kit 22221-524 is recommended for smaller muscle masses, such as mouse tibialis.

Electrodes may be used with electroporators 58018-004, 58017-932, and 58017-984. The ECM®/Enhancer series (82019-182) is recommended for monitoring the electrodes.

Electrode consists of a reusable Delrin® plastic handle and a sterile, disposable two-needle array assembly. Needle array assemblies are packaged in “six packs,” allowing for guaranteed sterility. Needles are made of medical-grade stainless steel.

Ordering information: Kit 22221-522 includes a 10mm diameter two-needle array handle and a sterile package of six electrode assemblies with 10mm spacing. Kit 22221-524 includes a 5mm diameter handle and a sterile package of six electrode assemblies with 5mm spacing.

Specifications
  • Description:
    2-Needle Array™, electrode gap 10 mm
  • Handle length:
    8 cm
  • Needle length:
    20 mm
  • Cat. No.:
    22221-522
  • Voltage range:
    0 - 500 V
  • Pulse length range:
    1 µs to 99 ms in PBS (Phosphate Buffer Saline)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.