Orders LinkContactUsLinkComponent
Optimize Back Pressure Regulators
Optimize Back Pressure Regulators
10 psi back pressure regulator
Catalog # 21510-684
Supplier:   Optimize
Supplier #:  10-06-00120
Optimize Back Pressure Regulators
Catalog # 21510-684
Supplier:  Optimize
Optimize Back Pressure Regulators
Optimize Back Pressure Regulators
10 psi back pressure regulator
Price
$888.46
Log in for availability
Each
Product Details & Documents
Optimize flow-through back pressure regulators utilize an active mechanism to maintain steady back pressure across a range of mobile phase viscosities and flow rates.
  • Flow-through design
  • Utilizes an active mechanism to maintain steady back pressure across a range of conditions
  • Prevent solvent decompression and bubble formation in the flow cell
  • Reduces baseline noise and drift
  • 100 µl internal volume
Placing one downstream from your detector will prevent solvent decompression and bubble formation in the flow cell, reducing baseline noise and drift. Each pre-set Optimize stainless steel Back Pressure Regulator offers a 100 µl internal volume and includes fittings for immediate connection out of the box.
Delivery information: Includes OPTI-LOK® LP Fittings.
Specifications
  • Description:
    10 psi back pressure regulator
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.