VWR® Micro Burets, Class A
Micro Buret, Class A, 1 ml
Catalog # 17477-020
Supplier: VWR International
Jump to:
Product Details & Documents
Borosilicate glass buret manufactured to Class A specifications for burets.
- Borosilicate glass fabrication
Specifications
- Capacity:1 ml
- Description:Micro Buret, Class A