Orders LinkContactUsLinkComponent
DicksonWare™ Software, Dickson
DicksonWare™ Software, Dickson
Secure Software with USB Download Cable
Catalog # 14905-016
DicksonWare™ Software, Dickson
Catalog # 14905-016
Supplier:  
DicksonWare™ Software, Dickson
DicksonWare™ Software, Dickson
Secure Software with USB Download Cable
Discontinued
Product Details & Documents
Software allows for simple viewing and zooming of logged data.
The software is also available in secure, 21CFR11-compliant version. This version provides password protection, data encryption, and an audit trail of user activity. Secure software labels all charts with the user ID and serial number of data logger used.
Software features user selectable sample intervals, delayed logger start times, and temperature displays in Celsius or Fahrenheit. Data and graphs can be downloaded quickly, exported effortlessly, and viewed in tabular or graphical formats. Allows for real-time monitoring. Microsoft Windows® compatible. Software includes USB or serial download cable.
Specifications
  • Description:
    Secure Software with USB Download Cable