Order Entry
Canada
Orders LinkContactUsLinkComponent
Corning® Pasteur Pipettes

Corning® Pasteur Pipettes

Disposable pasteur pipettes, Soda Lime Glass, 6.5 mm, 146.05 mm (5¾"), Non sterile
Catalog # 14673-156
Corning® Pasteur Pipettes
Catalog # 14673-156
Supplier:  
Corning® Pasteur Pipettes
Corning® Pasteur Pipettes

Disposable pasteur pipettes, Soda Lime Glass, 6.5 mm, 146.05 mm (5¾"), Non sterile

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PIPETTES PASTEUR 146MM CS1000 Catalog #53499-630

 

PIPET DISPOSABLE 5-3/4IN PK250 Catalog #KT883350-0575

 

Product Details & Documents

Soda-lime or borosilicate glass.

  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs

These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures.

Specifications
  • Ext.Ø:
    6.5 mm
  • Sterility:
    Non sterile
  • Description:
    Disposable pasteur pipettes
  • Length:
    146.05 mm (5¾")
  • Material:
    Soda Lime Glass