VWR®, Beaker Cups, Polystyrene
250 ml (8.5 oz.)
Catalog # 13916-060
Supplier: VWR International
Jump to:
Product Details & Documents
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
- Disposable
- Polystyrene fabrication
- Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
Documents
Certificates
Search by Lot Number13915-996_P 13916-004_P 13916-015_P 13916-037_P 13916-060_P
BSE/TSE Certificates
Specifications
- Capacity:250 ml (8.5 oz.)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



