Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Bath Fluids

VWR® Bath Fluids

PolyScience polytherm S250, 3.8 l (1 gal.), VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems, 150...250 °C (302...482 °F), (25°C) 500 cSt
Catalog # 13270-792
Supplier:   VWR International
VWR® Bath Fluids
Catalog # 13270-792
Supplier:  VWR International
CAS Number:  
VWR® Bath Fluids
VWR® Bath Fluids

PolyScience polytherm S250, 3.8 l (1 gal.), VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems, 150...250 °C (302...482 °F), (25°C) 500 cSt

CAS Number:  
Price
$3,876.06
New Customer Promotional Price
Log in for availability
Each (1US Gal)
Product Details & Documents

These heat-transfer fluids provide superior thermal stability for high or low temperatures.

  • Silicone heat transfer fluid

Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.

Packaging: Bottles are packaged in a carton for shipment.

Specifications

Specifications

  • Description:
    PolyScience polytherm S250
  • Size:
    3.8 l (1 gal.)
  • Temperature range:
    150...250 °C (302...482 °F)
  • For use with:
    VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems
  • Viscosity:
    (25°C) 500 cSt

Specifications