Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Modular Heating Sand Baths

VWR® Modular Heating Sand Baths

Sand Bath for 1 Block Heater, 9.5 x 7.6 x 6.4 cm (33/4 x 3 x 21/2")
Catalog # 13259-262
Supplier:   VWR International
VWR® Modular Heating Sand Baths
Catalog # 13259-262
Supplier:  VWR International
VWR® Modular Heating Sand Baths
VWR® Modular Heating Sand Baths

Sand Bath for 1 Block Heater, 9.5 x 7.6 x 6.4 cm (33/4 x 3 x 21/2")

Price
$931.85
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

For use in heating blocks, these sand baths are ideal for irregular vessels.

  • Now with Best-in-Class, Exclusive 5-Year Warranty

Stainless steel construction for superior corrosion resistance. Designed to hold sand, shot, or nonvolatile liquids. Right angle thermometers (61220-740 and 61220-742) have an accuracy rating of ± 1°C.

Documents

Specifications
  • Description:
    Sand Bath for 1 Block Heater
  • L×W×H:
    9.5 x 7.6 x 6.4 cm (33/4 x 3 x 21/2")
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stainless Steel Shot for Sand Bath, 454 g (1 lb.)
    Stainless Steel Shot for Sand Bath, 454 g (1 lb.)
    13259-274
    Log in for availability
    Each - $257.84