VWR® Talon® Support Stands, Cast Iron, Rectangular Base
15.2 x 22.9 cm (6 x 9")
Catalog # 12985-072
Supplier: VWR International
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
Catalog # 12985-072
Supplier: VWR International
Jump to:
Product Details & Documents
Support stand bases are constructed of cast iron with black enamel finish.
- Cast iron construction with black enamel finish
- Built-in support rod holder
- Rectangular base
- For use with VWR® Talon® lab frames
All bases feature a built-in support rod holder with locking knob that accepts a 13mm (1/2") diameter support rod (sold separately).
Documents
VWR Laboratory Equipments
Product Literature
Specifications
- Dimensions:15.2 x 22.9 cm (6 x 9")
Accessories
Product Information
- Catalog #ØMaterialRod LengthAvailabilityPrice
Product Information
- Catalog #ØMaterialRod LengthAvailabilityPrice
Product Information
- Catalog #ØMaterialAvailabilityPrice
- Support Rod
Product Information
- Catalog #ØMaterialAvailabilityPrice
- Support Rod
Product Information
- Catalog #ØMaterialAvailabilityPrice
- Support Rod
Product Information
- Catalog #ØMaterialAvailabilityPrice
- Support Rod
Product Information
- Catalog #ØMaterialRod LengthAvailabilityPrice
Product Information
- Catalog #ØMaterialRod LengthAvailabilityPrice