Orders LinkContactUsLinkComponent
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
12.7 x 20.3 cm (5 x 8")
Catalog # 12985-070
Supplier:   VWR International
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
Catalog # 12985-070
Supplier:  VWR International
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
12.7 x 20.3 cm (5 x 8")
Price
226,03 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Support stand bases are constructed of cast iron with black enamel finish.
  • Cast iron construction with black enamel finish
  • Built-in support rod holder
  • Rectangular base
  • For use with VWR® Talon® lab frames
All bases feature a built-in support rod holder with locking knob that accepts a 13mm (1/2") diameter support rod (sold separately).
Documents
Specifications
  • Dimensions:
    12.7 x 20.3 cm (5 x 8")
Accessories
Product Information
  • Catalog #
    Ø
    Material
    Rod Length
    Availability
    Price
  • 60079-360
    13 mm (1/2")
    Aluminium
    305 mm (12")
    Log in for availability
    Each - $60.74
Product Information
  • Catalog #
    Ø
    Material
    Rod Length
    Availability
    Price
  • 60079-428
    13 mm (1/2")
    Aluminium
    914 mm (36")
    Log in for availability
    Each - $74.92
Product Information
  • Catalog #
    Ø
    Material
    Availability
    Price
  • Support Rod
    Support Rod
    60079-533
    13 mm (1/2")
    Stainless steel
    Log in for availability
    Each - $61.60
Product Information
  • Catalog #
    Ø
    Material
    Availability
    Price
  • Support Rod
    Support Rod
    60079-534
    13 mm (1/2")
    Stainless steel
    Log in for availability
    Each - $86.31
Product Information
  • Catalog #
    Ø
    Material
    Availability
    Price
  • Support Rod
    Support Rod
    60079-535
    13 mm (1/2")
    Stainless steel
    Log in for availability
    Each - $96.60
Product Information
  • Catalog #
    Ø
    Material
    Availability
    Price
  • Support Rod
    Support Rod
    60079-536
    13 mm (1/2")
    Stainless steel
    Log in for availability
    Each - $140.54
Product Information
  • Catalog #
    Ø
    Material
    Rod Length
    Availability
    Price
  • 60079-381
    13 mm (1/2")
    Aluminium
    457 mm (18")
    Log in for availability
    Each - $64.17
Product Information
  • Catalog #
    Ø
    Material
    Rod Length
    Availability
    Price
  • 60079-406
    13 mm (1/2")
    Aluminium
    610 mm (24")
    Log in for availability
    Each - $70.11