Orders LinkContactUsLinkComponent
Troemner Alloy 8 Precision Analytical Weight Sets, Class 3, Troemner
17, 100 g - 10 mg
Catalog # 12622-251
Supplier:   Troemner
 :  80859292
Troemner Alloy 8 Precision Analytical Weight Sets, Class 3, Troemner
Catalog # 12622-251
Supplier:  Troemner
Troemner Alloy 8 Precision Analytical Weight Sets, Class 3, Troemner
Troemner Alloy 8 Precision Analytical Weight Sets, Class 3, Troemner
17, 100 g - 10 mg
Price
$3,408.08
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Highly polished kilogram and gram weights made of Troemner Alloy 8, a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are made of aluminum); fractional weights 3mg and below are made of aluminum. Weights 500 g and smaller are supplied in rugged polypropylene cases. Sets that include weights 1 kg and larger are supplied in oak, aluminum, or rugged plastic cases. All cases are fitted with individual compartments and marked with the accuracy class and serial number. A clear fitted cover secures the fractional weights within the case. Each case includes contoured weight forceps. Analytical weights 1g and above are manufactured with adjusting cavities.
This exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5°C (–320°F).
Certifications: Standard weight sets are supplied with a Statement of Accuracy. Where applicable, weight sets are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWR[email protected] for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
Specifications
  • Certification:
    NVLAP
  • Weight:
    100 g - 10 mg
  • No. of weights:
    17