Trigger Spray Bottles, Qorpak
473 ml (16 oz.), Nylon/PE, Modern Round, 28-400, White PP spray/stream trigger sprayer
Catalog # 11311-616
Supplier: Qorpak
Supplier #: PLC-06934
Jump to:
Product Details & Documents
Trigger spray bottles feature a trigger-motion spray that dispenses a spray or stream of liquid from an adjustable nozzle.
The translucent HDPE bottles are ideal for general use. HDPE provides an excellent moisture barrier and impact resistance. The adjustable sprayer permits a versatile spray pattern, from fine mist to coarse spray to jet stream.
Documents
Qorpak Laboratory Container and Supply
Product Literature
Specifications
- Cap Size:28-400
- Cap type:White PP spray/stream trigger sprayer
- Capacity:473 ml (16 oz.)
- Description:Modern Round
- Material:Nylon/PE
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



