Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Sample Vials, Clear Borosilicate Glass, Screw-Thread

Vials without treatment, 1.85 ml, 14.65×26 mm, Clear, No, 13-425
Catalog # 11311-218
Supplier:   Qorpak
 :  GLA-07796
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Catalog # 11311-218
Supplier:  Qorpak
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Sample Vials, Clear Borosilicate Glass, Screw-Thread

Vials without treatment, 1.85 ml, 14.65×26 mm, Clear, No, 13-425

Price
$367.72
New Customer Promotional Price
Log in for availability
Case of 144
Product Details & Documents

Clear, flint screw-thread sample vials offer maximum visibility.

  • Available with a variety of caps, or as vials only, to meet specific requirements
  • Pre-cleaned options save time and money

These general-purpose glass vials are made of Type I Borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.

Specifications
  • Cap included:
    No
  • Cap Size:
    13-425
  • Capacity:
    1.85 ml
  • Color:
    Clear
  • Description:
    Vials without treatment
  • Diameter ×H:
    14.65×26 mm