$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
SERVICE VALIDATION TOOL KIT
Catalog # 11010-450
Supplier:
QLA
Expand image
SERVICE VALIDATION TOOL KIT
Catalog # 11010-450
Supplier:
QLA
Supplier Number:
RECAL-KT
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH