Orders LinkContactUsLinkComponent
Disintegration Testing, Quality Lab Accessories
Clear Glass Disintegration Beaker with Flat Top, 900 mL
Catalog # 11010-360
Supplier:   QLA
 :  GLA905-DB
Disintegration Testing, Quality Lab Accessories
Catalog # 11010-360
Supplier:  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Clear Glass Disintegration Beaker with Flat Top, 900 mL
Price
$307.04
Log in for availability
Each
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Specifications
  • Description:
    Clear Glass Disintegration Beaker with Flat Top, 900 mL