Orders LinkContactUsLinkComponent
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
Catalog # 11010-218
Supplier:   QLA
Supplier #:  DISASY-06
Disintegration Testing, Quality Lab Accessories
Catalog # 11010-218
Supplier:  QLA
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
Price
$1,339.37
Log in for availability
Each
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Specifications
  • Description:
    Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers