Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Phosphate Buffered Saline, 1X, pH 7.4, Teknova

1X PBS Solution, Sterile, 1000 mL, 7.4
Catalog # 101449-092
Supplier:   Teknova
 :  P0261
Phosphate Buffered Saline, 1X, pH 7.4, Teknova
Catalog # 101449-092
Supplier:  Teknova
CAS Number:  
Phosphate Buffered Saline, 1X, pH 7.4, Teknova

1X PBS Solution, Sterile, 1000 mL, 7.4

CAS Number:  
Price
$318.04
Log in for availability
Each (1,000ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

pH adjusted to 7.4

  • Monobasic
  • Shipping Conditions: Ambient
  • Storage Conditions: Room Temperature

Documents

Specifications

Specifications

  • Description:
    1X PBS Solution, Sterile
  • Size:
    1000 mL
  • pH:
    7.4

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.