Orders LinkContactUsLinkComponent
Ampicillin
5 g
  MPCAAAJ60977-06
New Product
 :  J60977-06
Ampicillin
  MPCAAAJ60977-06
New Product
 :  69-53-4
Ampicillin
5 g
 :  69-53-4
$170.65
Each
Powder
CAS No.: 69-53-4 Molecular Formula: C16H19N3O4S Formula Weight: 349.40 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 208° dec. Solubility: In 1 M NH4OH, 50 mg/ml Beilstein Reference: 1090925 Merck Reference: 14,586 MDL No.: MFCD00005175
  •  :69-53-4
  •  :C₁₆H₁₉N₃O₄S
  •  :MFCD00005175
  •  :349.4 g/mol
  •  :14,586
  •  :208 °C (dec.)
  •  :Refrigerator
  •  :200-709-7
  • Size:
    5 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.