Order Entry
Canada
Orders LinkContactUsLinkComponent
Mops Buffer, 10X, Teknova

Mops Buffer, 10X, Teknova

10X Mops buffer, sterile solution, 100 ml
Catalog # CA77443-134
New Product
Supplier:   Teknova
Supplier #:  M2101
Mops Buffer, 10X, Teknova
Catalog # CA77443-134
Supplier:  Teknova
New Product
CAS Number:  
Mops Buffer, 10X, Teknova

10X Mops buffer, sterile solution, 100 ml

CAS Number:  
Price
$107.09
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

10X MOPS Buffer for EZ Rich defined medium kit

Repeated freeze thaw causes precipitation and a reduction in cell growth. Aliquot contents upon intial thaw

Suitable for molecular biology, protein chemistry, and biochemical applications.

Specifications

Specifications

  • Description:
    10X Mops buffer, sterile solution
  • Size:
    100 ml
  • Cat. No.:
    CA77443-134
  • Storage temperature:
    Frozen
  • Shipping temperature:
    Frozen

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.