Orders LinkContactUsLinkComponent
First Strand Buffer, 5X, Teknova
First strand buffer, 5X, 500 ml
Catalog # CA77443-420
New Product
Supplier:   Teknova
 :  F1204
First Strand Buffer, 5X, Teknova
Catalog # CA77443-420
Supplier:  Teknova
New Product
CAS Number:  
First Strand Buffer, 5X, Teknova
First strand buffer, 5X, 500 ml
CAS Number:  
Price
$313.49
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
First strand synthesis buffer designed for use in reverse transcription.
Consists of Tris-HCl (250 mM), pH 8.3, potassium chloride (375 mM), magnesium chloride (15 mM) and DTT (50 mM).
Specifications
Specifications
  • Description:
    First strand buffer, 5X
  • Size:
    500 ml
Specifications