Ampicillin solution 100 mg/mL
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Ampicillin is used as a selectable marker in many commonly used plasmids. A functional Ampicillin gene will confer resistance to cells containing the plasmid.
- Inhibits cell wall synthesis by inhibiting formation of the peptidoglycan cross-linkages
- Acts as a competitive inhibitor of transpeptidase
Inhibits bacterial cell wall synthesis by binding to one or more of the penicillin binding proteins (PBPs) that inhibit the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls, thus inhibiting cell wall biosynthesis.
- CAS Number:69-53-4
- Formula:C₁₆H₁₉N₃O₄S
- MDL Number:MFCD00005175
- Molecular Weight:349.4 g/mol
Specifications
Specifications
- Size:25 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



