Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Technik™ Chain Nose Pliers

Extra Long Chain Nose Pliers, 12.7 cm (5"), 3.7 cm (1⁷⁄₁₆″), Serrated
Catalog # 78621-012
New Product
Supplier:   Aven Tools
 :  10311S
Technik™ Chain Nose Pliers
Catalog # 78621-012
Supplier:  Aven Tools
New Product
Technik™ Chain Nose Pliers
Technik™ Chain Nose Pliers

Extra Long Chain Nose Pliers, 12.7 cm (5"), 3.7 cm (1⁷⁄₁₆″), Serrated

Price
$38.17
Log in for availability
Each
Product Details & Documents

Pliers feature dual leaf springs and box joint construction for comfortable action.

  • Made of mirror polished stainless steel
  • Induction hardened cutting edges
  • ESD-safe grips
Specifications
  • Description:
    Extra Long Chain Nose Pliers
  • Length:
    12.7 cm (5")
  • Jaw type:
    Serrated
  • Jaw length:
    3.7 cm (1⁷⁄₁₆″)