Order Entry
Canada
Orders LinkContactUsLinkComponent
Nitrate Test Kits

Nitrate Test Kits

NED, 0 - 210 ppm, 30
Catalog # 78620-298
New Product
Supplier:   CHEMetrics
Supplier #:  R-6901
Nitrate Test Kits
Catalog # 78620-298
Supplier:  CHEMetrics
New Product
CAS Number:  
Nitrate Test Kits
Nitrate Test Kits

NED, 0 - 210 ppm, 30

CAS Number:  
Price
$98.04
Log in for availability
Pack of 30
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The NED method is widely used in the quantitative analysis of nitrite and nitrate in water samples. NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, lower detection limits and measurement ranges are possible vs. chromotropic acid kits. In addition, the nitrate measurement uses only zinc which is less toxic than cadmium, making handling and disposal safer.

  • Measurement via zinc reduction only, safer handling and disposal

    Improved sensitivity and faster analysis time

    Simplified reordering, visual refills include NED, Zn, Acidifier and buffer under single catalog code

CHEMetrics Nitrate Test Kits use the N-(1-Naphthyl) ethylenediamine (NED) method to measure nitrate in potable, surface, storm, ground, sea, and wastewater. These kits offer improved sensitivity and faster analysis time (10 vs. 13 min) compared to our current chromotropic acid-based kits.

Like our legacy chromotropic acid reagent, NED readily undergoes a diazonium coupling reaction in the presence of nitrite to give a strongly colored pink azo compound. The intensity of the color is directly proportional to the concentration of nitrite in the sample. Due to the significant intensity of the developed color even at very low nitrite/nitrate concentrations, we are now able to offer lower detection limits and measurement ranges than our chromotropic acid kits provide.

Specifications

Specifications

  • Chemical method:
    NED
  • Application:
    Drinking water; watershed and ecosystem health; wastewater; aquaculture; agriculture runoff
  • Range:
    0 - 210 ppm
  • No. of tests:
    30

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.