Orders LinkContactUsLinkComponent
Schiff reagent modified | Avantor
Schiff reagent modified
Glass bottle, 500 ml
Catalog # CA78612-036
New Product
Supplier:   CHAPTEC INC CA
Fournisseur #:  RSY2250-500
Schiff reagent modified
Catalog # CA78612-036
Supplier:  CHAPTEC INC CA
New Product
CAS Number:  7647-01-0; 569-61-9
Schiff reagent  modified
Schiff reagent modified
Glass bottle, 500 ml
CAS Number:  7647-01-0; 569-61-9
Price
$51.82
Log in for availability
Each(1/Itm/Bx)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Schiff's Reagent is a ready-to-use solution used during P.A.S (Periodic Acid, Schiff) staining. It is used for the detection of aldehydes and mucopolysaccharides, glycogen, muco- and glycoproteins, glycolipids, phospholipids, basement membranes and collagen in histology.
  • Reacts with aldehydes to form a bright red color
  • Widely used for the detection of a certain functional group in chemical solutions
  • This modified solution is similar to Sigma's formulation
  • CAS Number:7647-01-0; 569-61-9
Specifications
Specifications
  • Pack type:
    Glass bottle
  • Size:
    500 ml
Specifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.