Schiff reagent modified
Glass bottle, 1 L
Catalog # CA78612-030
New Product
Supplier: CHAPTEC INC CA
Supplier #: RSY2250-1
Schiff reagent modified
Catalog # CA78612-030
Supplier: CHAPTEC INC CA
New Product
CAS Number: 7647-01-0; 569-61-9
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Schiff's Reagent is a ready-to-use solution used during P.A.S (Periodic Acid, Schiff) staining. It is used for the detection of aldehydes and mucopolysaccharides, glycogen, muco- and glycoproteins, glycolipids, phospholipids, basement membranes and collagen in histology.
Reacts with aldehydes to form a bright red color
- Widely used for the detection of a certain functional group in chemical solutions
- This modified solution is similar to Sigma's formulation
- CAS Number:7647-01-0; 569-61-9
Specifications
Specifications
- Pack type:Glass bottle
- Size:1 L