Deolorizing Solutions 1%
Decolorizing solution 1%, 500 ml, Plastic bottle
Catalog # CA78612-020
New Product
Supplier: CHAPTEC INC CA
Supplier #: DEY0100-500
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Decolorizing solution for H&E & PAP stain.
- Available in various concentrations of hydrochloric acid (0.25%, 0.5%, and 1%) in 70% alcohol
The 1% alcohol-acid differentiation solution is a 70% alcoholic solution containing 1% concentrated hydrochloric acid. It is commonly used in histology and cytology to remove excess stain and highlight nuclei after hematoxylin staining in the H&E and PAP methods. To achieve H&E staining with precise nuclear details, it is crucial that certain hematoxylins are properly differentiated, making the alcohol-acid solution particularly effective.
Specifications
Specifications
- Pack type:Plastic bottle
- Description:Decolorizing solution 1%
- Application:Destaining reagents
- Volume:500 ml
- Cat. No.:CA78612-020
- CAS No.:64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



