Order Entry
Canada
Orders LinkContactUsLinkComponent
Deolorizing Solutions 1%

Deolorizing Solutions 1%

Decolorizing solution 1%, 1 L, Plastic bottle
Catalog # CA78612-134
New Product
Supplier:   CHAPTEC INC CA
Supplier #:  DEY0100-1
Deolorizing Solutions 1%
Catalog # CA78612-134
Supplier:  CHAPTEC INC CA
New Product
CAS Number:  
Deolorizing Solutions 1%
Deolorizing Solutions 1%

Decolorizing solution 1%, 1 L, Plastic bottle

CAS Number:  
Price
$29.76
Log in for availability
Each(1/Itm/Bx)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Decolorizing solution for H&E & PAP stain.

  • Available in various concentrations of hydrochloric acid (0.25%, 0.5%, and 1%) in 70% alcohol

The 1% alcohol-acid differentiation solution is a 70% alcoholic solution containing 1% concentrated hydrochloric acid. It is commonly used in histology and cytology to remove excess stain and highlight nuclei after hematoxylin staining in the H&E and PAP methods. To achieve H&E staining with precise nuclear details, it is crucial that certain hematoxylins are properly differentiated, making the alcohol-acid solution particularly effective.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Description:
    Decolorizing solution 1%
  • Application:
    Destaining reagents
  • Volume:
    1 L
  • Cat. No.:
    CA78612-134
  • CAS No.:
    64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.