Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Deolorizing Solutions 0.25%

Decolorizing solution 0.25%, 4 L, Plastic jug
Catalog # CA78612-088
New Product
Supplier:   CHAPTEC INC CA
 :  DEY0025
Deolorizing Solutions 0.25%
Catalog # CA78612-088
Supplier:  CHAPTEC INC CA
New Product
CAS Number:  
Deolorizing Solutions 0.25%
Deolorizing Solutions 0.25%

Decolorizing solution 0.25%, 4 L, Plastic jug

CAS Number:  
Price
$123.10
Log in for availability
Each(1/Itm/Bx)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Decolorizing solution for H&E & PAP stain.

  • Available in various concentrations of hydrochloric acid (0.25%, 0.5%, and 1%) in 70% alcohol

The 0.25% alcohol-acid differentiation solution is a 70% alcoholic solution containing 0.25% concentrated hydrochloric acid. It is commonly used in histology and cytology to remove excess stain and highlight nuclei after hematoxylin staining in the H&E and PAP methods. To achieve H&E staining with precise nuclear details, it is crucial that certain hematoxylins are properly differentiated, making the alcohol-acid solution particularly effective.

Specifications

Specifications

  • Pack type:
    Plastic jug
  • Description:
    Decolorizing solution 0.25%
  • Application:
    Destaining reagents
  • Volume:
    4 L
  • Cat. No.:
    CA78612-088
  • CAS No.:
    64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)

Specifications