Deolorizing Solutions 0.5%
Decolorizing solution 0.5%, 1 L, Plastic bottle
Catalog # CA78612-204
New Product
Supplier: CHAPTEC INC CA
Supplier #: DEY0050-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Decolorizing solution for H&E & PAP stain.
- Available in various concentrations of hydrochloric acid (0.25%, 0.5%, and 1%) in 70% alcohol
The 0.5% alcohol-acid differentiation solution is a 70% alcoholic solution containing 0.5% concentrated hydrochloric acid. It is commonly used in histology and cytology to remove excess stain and highlight nuclei after hematoxylin staining in the H&E and PAP methods. To achieve H&E staining with precise nuclear details, it is crucial that certain hematoxylins are properly differentiated, making the alcohol-acid solution particularly effective.
Specifications
Specifications
- Pack type:Plastic bottle
- Description:Decolorizing solution 0.5%
- Application:Destaining reagents
- Volume:1 L
- Cat. No.:CA78612-204
- CAS No.:64-17-5 (30.0 - 60.0%); 67-56-1 (7.0 - 13.0%); 141-78-6 (0.1 - 1.0%); 7647-01-0 (0.1 - 1.0%)



