Orders LinkContactUsLinkComponent
NEBExpress® T4 Lysozyme
NEBExpress® T4 Lysozyme
NEBExpress® T4 Lysozyme, 1 mg/ml, 1000 µg
Catalog # CA76717-038
New Product
Supplier #:  P8115L
NEBExpress® T4 Lysozyme
Catalog # CA76717-038
New Product
NEBExpress® T4 Lysozyme
NEBExpress® T4 Lysozyme
NEBExpress® T4 Lysozyme, 1 mg/ml, 1000 µg
Price
$658.00
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
NEBExpress T4 Lysozyme is 200-fold more active than chicken egg white lysozyme.
  • Lysis reactions are scalable and compatible with high throughput workflows
  • Fast non-mechanical bacterial lysis; the lysate is ready to use and compatible with affinity resins
  • Small (200 µg) and large (5×200 µg) formats suitable for lysing 7 or 35 g of cells, respectively
  • Recombinant, animal free and REACH compliant
NEBExpress T4 Lysozyme is a recombinant murein hydrolase that breaks down the bacterial cell wall by hydrolyzing the beta 1 to 4 linkage between N-acetylmuramic acid and N-acetylglucosamine in the peptidoglycan of prokaryotic cells (gram-negative and some gram-positive bacteria). It can be used to extract soluble proteins, membrane proteins, DNA, RNA, or metabolites.
Specifications
  • Description:
    NEBExpress® T4 Lysozyme, 1 mg/ml
  • Size:
    1000 µg