Transferpette® pro, Ejector Set (upper and lower)
1 - 10 ml
Catalog # 78599-788
New Product
Supplier: Brandtech
Supplier #: 704757
Transferpette® pro, Ejector Set (upper and lower)
Catalog # 78599-788
Supplier: Brandtech
New Product
Jump to:
Product Details & Documents
Replacement ejector set.
Ejector set (upper and lower part) for use with Transferpette® pro single channel pipettors, 2.5 to 10ml
Designed for a perfect fit, simple installation, and long-lasting reliability.
Specifications
- Volume:1 - 10 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



