Orders LinkContactUsLinkComponent
Transferpette® pro, Ejector (bottom part) | Avantor
Transferpette® pro, Ejector (bottom part)
2 - 20 µl
Catalog # 78599-756
New Product
Supplier:   Brandtech
Fournisseur #:  704710
Transferpette® pro, Ejector (bottom part)
Catalog # 78599-756
Supplier:  Brandtech
New Product
Transferpette® pro, Ejector (bottom part)
Transferpette® pro, Ejector (bottom part)
2 - 20 µl
Price
$43.83
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Replacement lower ejector.
  • Ejector (bottom part) for use with Transferpette® pro single channel pipettors, up to 1,000µl
Designed for a perfect fit, simple installation, and long-lasting reliability.
Specifications
  • Volume:
    2 - 20 µl
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.