Orders LinkContactUsLinkComponent
Hanna Instruments NaCl Solution
30 g/L NaCl Standard Solution (230 ml)
Catalog # CA470122-054
New Product
Supplier:   Hanna
 :  HI7081M
Hanna Instruments NaCl Solution
Catalog # CA470122-054
Supplier:  Hanna
New Product
CAS Number:  
Hanna Instruments NaCl Solution
Hanna Instruments NaCl Solution
30 g/L NaCl Standard Solution (230 ml)
CAS Number:  
Price
$26.03
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This premium quality 30 g/L NaCl standard solution for the calibration of salinity meters and sodium ISEs. Hanna calibration solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna's line of calibration solutions have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.
  • 29 g/L NaCl standard solution
  • Air tight bottle with tamper-proof seal to ensure quality
This premium quality 30 g/L NaCl calibration standard solution produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers. Reported values are traceable to NIST Standard Reference Materials (SRMs).
Specifications
Specifications
  • Description:
    30 g/L NaCl Standard Solution (230 ml)
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)