Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Hanna Instruments Zero Oxygen Solution Set (500 ml)

Zero oxygen solution set (500 ml)
  CA470001-918
New Product
 :   Hanna
 :  HI7040L
Hanna Instruments Zero Oxygen Solution Set (500 ml)
  CA470001-918
 :  Hanna
New Product
 :  
Hanna Instruments Zero Oxygen Solution Set (500 ml)
Hanna Instruments Zero Oxygen Solution Set (500 ml)

Zero oxygen solution set (500 ml)

 :  
 
$27.47
   
Each
 
   
 

This is a high quality, bicomponent zero oxygen solution made from reagent grade chemicals. Hanna dissolved oxygen solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution.

  • Prepared with reagent grade chemicals
  • Hassle-free, easy to mix chemical component bottles
  • Air-tight seal to ensure the quality of the solution

This solution is a bicomponent zero oxygen solution designed for calibrating both polarographic and galvanic dissolved oxygen sensors. By simply immersing the dissolved oxygen probe in the solution users can quickly calibrate to 0% oxygen concentration, allowing for accurate follow-up measurements of dissolved oxygen in their sample. Hanna dissolved oxygen solutions are produced in our state-of-the-art manufacturing facility using reagent grade chemicals in a temperature controlled environment. 

 

 

 

  • Description:
    Zero oxygen solution set (500 ml)

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.