Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Optifit Pipette Tips

10 ml, 164 mm, Transparent, Red, Sartorius pipettes, 1 bag
Catalog # 78563-824
New Product
Supplier:   Sartorius
 :  LH-799004
Optifit Pipette Tips
Catalog # 78563-824
Supplier:  Sartorius
New Product
Optifit Pipette Tips
Optifit Pipette Tips

10 ml, 164 mm, Transparent, Red, Sartorius pipettes, 1 bag

Price
$76.70
Log in for availability
Pack of 210
Product Details & Documents

The non-filtered Optifit tips are designed to seamlessly fit together with Sartorius pipettes. For applications involving larger volume transfers, the 10 ml Optifit tips are specifically designed to deliver precise and accurate performance.

  • Informative rack labels indicating tip volume, product number, lot number, expiration date, and purity level for improved tip identification and traceability

The Optifit tips are intended, designed and manufactured to be used in liquid dispensing in a variety of applications and to be used in combination with Sartorius pipettes.

Specifications
  • Volume:
    10 ml
  • Length:
    164 mm
  • Tip color:
    Transparent
  • Packed:
    1 bag
  • For use with:
    Sartorius pipettes
  • Color code:
    Red