Orders LinkContactUsLinkComponent
Exploding Bomb - GHS Pictogram Labels
Exploding Bomb - GHS Pictogram Labels
Durable adhesive vinyl, 4×4", 25/pack
Catalog # 78542-962
New Product
Supplier:   HCL LABELS, INC
Supplier #:  SSL-1090-0044
Exploding Bomb - GHS Pictogram Labels
Catalog # 78542-962
Supplier:  HCL LABELS, INC
New Product
Exploding Bomb - GHS Pictogram Labels
Exploding Bomb - GHS Pictogram Labels
Durable adhesive vinyl, 4×4", 25/pack
Price
$73.85
Log in for availability
Pack of 25
Product Details & Documents
OSHA/GHS-compliant exploding bomb hazard labels identify explosives, self-reactives, and organic peroxides. Available in durable vinyl, they are UV and chemical-resist. These labels ensure workplace safety and regulatory compliance.
  • Durable adhesive vinyl
  • Chemical-resistant
  • UV resistant
Specifications
  • Dimensions:
    4×4"
  • Material:
    Durable adhesive vinyl
  • Packaging:
    25/pack