Tapestri® Software
Tapestri® Single-Cell Genome Integrity CNV Software, portal access, Annual access to Tapestri® Single-Cell Genome Integrity CNV analysis software hosted on portal.
78542-226
New Product
: MB06-0012
Access to the cloud analysis pipeline for data processing is included. For additional tertiary analysis and specialized reporting, we offer user friendly software. Pipelines specialized in the analysis of Genome Editing, Multiple Myeloma, and Genome Integrity.
: Does not include instrument, reagents or materials for processing samples. Access is provided to all laboratory users and starts on the order date.
- Description:Tapestri® Single-Cell Genome Integrity CNV Software, portal access
- Includes:Annual access to Tapestri® Single-Cell Genome Integrity CNV analysis software hosted on portal.