Orders LinkContactUsLinkComponent
Stopper Assembly
Stopper Assembly
Stopper assembly, #6 taper, 30 mm
Catalog # 78473-510
New Product
Supplier:   SAVILLEX
Supplier #:  730-0066
Stopper Assembly
Catalog # 78473-510
Supplier:  SAVILLEX
New Product
Stopper Assembly
Stopper assembly, #6 taper, 30 mm
Price
125,56 $
Log in for availability
Each
Product Details & Documents
  • For use in glass flasks
    Ferrule nut accepts ³/₈" OD tube
Specifications
  • Description:
    Stopper assembly, #6 taper
  • Size:
    30 mm