Column Component Closures
Transfer closure, pressure relief valve, Ultem® clamp, 1¹/₂" FNPT threads, 58 mm
Catalog # 78474-575
New Product
Supplier: SAVILLEX
Supplier #: 600-058-87
Jump to:
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Specifications
- Description:Transfer closure, pressure relief valve, Ultem® clamp, 1¹/₂" FNPT threads, 58 mm