Column Component Closures
Column component closure, with two ¹/₈" push in transfer side ports, 24 mm
Catalog # 78473-842
New Product
Supplier: SAVILLEX
Supplier #: 600-024-23
Jump to:
Product Details & Documents
Closures are molded from PFA, offering a service temperature range of −200 to +260 °C, and are interchangeable for use with column components or segments; a wrench set is recommended for secure tightening.
Specifications
- Description:Column component closure, with two ¹/₈" push in transfer side ports, 24 mm