Mining Sampling Bags, Clear Poly
Poly sample bag, 6 mil, 12×18", Plastic, Clear
Catalog # 80702-042
New Product
Supplier: SELENE PLASTIC BAGS CA
Supplier #: 1218500500CL
Jump to:
Product Details & Documents
Clear poly, mining sampling bags in various sizes and mil thicknesses.
Suitable for field and lab applications
- Made in Canada
Side welded seal imparts improved, with greater strength, and a resulting solid tubing bottom edge.
Specifications
- Color:Clear
- Description:Poly sample bag
- Dimensions:12×18"
- Material:Plastic
- Thickness:6 mil