Orders LinkContactUsLinkComponent
Mining Sampling Bags, Clear Poly
Mining Sampling Bags, Clear Poly
Poly sample bag, 4 mil, 8×14", Plastic, Clear
Catalog # 80701-958
New Product
Supplier #:  0814391000CL
Mining Sampling Bags, Clear Poly
Catalog # 80701-958
New Product
Mining Sampling Bags, Clear Poly
Poly sample bag, 4 mil, 8×14", Plastic, Clear
Price
$214.35
Log in for availability
Case of 1,000
Product Details & Documents
Clear poly, mining sampling bags in various sizes and mil thicknesses.
  • Suitable for field and lab applications
  • Made in Canada
Side welded seal imparts improved, with greater strength, and a resulting solid tubing bottom edge.
Specifications
  • Color:
    Clear
  • Description:
    Poly sample bag
  • Dimensions:
    8×14"
  • Material:
    Plastic
  • Thickness:
    4 mil