Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Microbial Count Plate

Pivot Coliform count plate, Blue purple colony, 36±1 °C, 24±2h incubation
Catalog # 78172-270
New Product
 :  MCPG00425
Microbial Count Plate
Catalog # 78172-270
New Product
CAS Number:  
Microbial Count Plate

Pivot Coliform count plate, Blue purple colony, 36±1 °C, 24±2h incubation

CAS Number:  
Price
$349.35
Log in for availability
Pack of 25
Product Details & Documents

GVS ready-to-use microbial count plate for the enumeration of multiple microbials such as Aerobic bacteria, E. coli, yeast and molds.

  • Less time and less labor work ISO and AOAC standard compatible No more culture medium preparation No extra reagent required

GVS microbiology count plate based on selective medium to enumerate the food microorganisms in various samples.

Specifications

Specifications

  • Description:
    Pivot Coliform count plate
  • Format:
    Blue purple colony, 36±1 °C, 24±2h incubation

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.